3-Ethyl-(-)-thiazolidine-3,4-dicarboxylate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: (-)-3-Carbethoxythiazolidine-4-carboxylic Acid, (-)-3-Ethoxycarbonylthiazolidine-4-carboxylic Acid, (-)-Thiazolidine-3,4-dicarboxylic Acid 3-Ethyl Ester

T1974-25G T1974-5G
TCIAT1974-25GEA 196 EUR
TCIAT1974-25G TCIAT1974-5G
3-Ethyl-(-)-thiazolidine-3,4-dicarboxylate ≥98.0% (by titrimetric analysis)
3-Ethyl-(-)-thiazolidine-3,4-dicarboxylate
Formula: C₇H₁₁NO₄S
MW: 205,23 g/mol
Número MDL: MFCD04117921
Núm. CAS: 127657-29-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 97.0 to 100.0 deg-C
Specific rotation [a]20/D -115.0 to -125.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR