2,3,4,6-Tetra-O-benzoyl-D-glucopyranose ≥90.0% (by HPLC)

Proveedor: TCI

Sinónimos: 2,3,4,6-Tetrabenzoyl-D-glucopyranose

T2020-1G T2020-250MG
TCIAT2020-1GEA 399 EUR
TCIAT2020-1G TCIAT2020-250MG
2,3,4,6-Tetra-O-benzoyl-D-glucopyranose ≥90.0% (by HPLC)
2,3,4,6-Tetra-O-benzoyl-D-glucopyranose
Formula: C₃₄H₂₈O₁₀
MW: 596,59 g/mol
Número MDL: MFCD00066004
Núm. CAS: 64768-20-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 90.0 area%
Melting point 120.0 to 124.0 deg-C
Specific rotation [a]20/D +72.0 to +77.0 deg (C=1,Dioxane)
Molar extinction coefficient min. 43000 (5mg/L、MeOH、228.0 to 231.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR