1,3,4,6-Tetrakis(methoxymethyl)glycoluril ≥98.0% (by total nitrogen basis)

Proveedor: TCI

Sinónimos: 1,3,4,6-Tetrakis(methoxymethyl)tetrahydroimidazo[4,5-d]imidazole-2,5(1H,3H)-dione

T2058-25G T2058-5G
TCIAT2058-25GEA 98 EUR
TCIAT2058-25G TCIAT2058-5G
1,3,4,6-Tetrakis(methoxymethyl)glycoluril ≥98.0% (by total nitrogen basis)
1,3,4,6-Tetrakis(methoxymethyl)glycoluril
Formula: C₁₂H₂₂N₄O₆
MW: 318,33 g/mol
Número MDL: MFCD00402438
Núm. CAS: 17464-88-9
EINECS: 241-480-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 117.0 to 122.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR