Tris(dibenzylideneacetone)dipalladium(0) ≥75.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Pd2dba3, Pd₂(dba)₃
T2184-1G
T2184-5G
TCIAT2184-1GEA
178
EUR
TCIAT2184-1G
TCIAT2184-5G
Tris(dibenzylideneacetone)dipalladium(0) ≥75.0% (by titrimetric analysis)
Tris(dibenzylideneacetone)dipalladium(0)
Formula:
C₅₁H₄₂O₃Pd₂ MW: 915,72 g/mol |
Número MDL:
MFCD00013310 Núm. CAS: 51364-51-3 EINECS: 479-280-5 |
Resultados test especificaciones
Appearance | Red to Dark blue to Black powder to crystal |
Purity (Chelometric Titration) | min. 75.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...