4,4',4''-Tri-9-carbazolyltriphenylamine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
Proveedor: TCI
|
Atención
|
Sinónimos:
Tris(4-carbazol-9-ylphenyl)amine, 4,4',4''-Tris(carbazol-9-yl)triphenylamine, Tris(4-(9H-carbazol-9-yl)phenyl)amine, TCTA
T2274-200MG
T2274-1G
TCIAT2274-200MGEA
50
EUR
TCIAT2274-200MG
TCIAT2274-1G
4,4',4''-Tri-9-carbazolyltriphenylamine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
4,4',4''-Tri-9-carbazolyltriphenylamine
Formula:
C₅₄H₃₆N₄ MW: 740.91 g/mol Pto. de fusión: 266°C |
Número MDL:
MFCD03093250 Núm. CAS: 139092-78-7 |
|
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...