2,3,4,6-Tetra-O-acetyl-ɑ-D-galactopyranosyl-2,2,2-trichloroacetimidate ≥95.0% (by HPLC)
Proveedor: TCI
Sinónimos:
(2R,3S,4S,5R,6R)-2-(Acetoxymethyl)-6-(2,2,2-trichloro-1-iminoethoxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate
T2295-1G
T2295-5G
TCIAT2295-1GEA
105
EUR
TCIAT2295-1G
TCIAT2295-5G
2,3,4,6-Tetra-O-acetyl-ɑ-D-galactopyranosyl-2,2,2-trichloroacetimidate ≥95.0% (by HPLC)
2,3,4,6-Tetra-O-acetyl-alpha-D-galactopyranosyl-2,2,2-trichloroacetimidate
Formula:
C₁₆H₂₀Cl₃NO₁₀ MW: 492,68 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD07369652 Núm. CAS: 86520-63-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Melting point | 120.0 to 124.0 deg-C |
Specific rotation [a]20/D | +109 to +115 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...