2,3,4,5-Tetrachlorobenzoic acid ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Atención

T2378-25G T2378-5G
TCIAT2378-25GEA 60 EUR
TCIAT2378-25G TCIAT2378-5G
2,3,4,5-Tetrachlorobenzoic acid ≥97.0% (by GC, titration analysis)
2,3,4,5-Tetrachlorobenzoic acid
Formula: C₇H₂Cl₄O₂
MW: 259,89 g/mol
Número MDL: MFCD00061126
Núm. CAS: 50-74-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 193.0 to 197.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR