(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: H-D-Trp-OMe·HCl, D-Tryptophan methyl ester hydrochloride, H-D-Trp-OMe.HCl

T2442-25G T2442-5G
TCIAT2442-25GEA 92 EUR
TCIAT2442-25G TCIAT2442-5G
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride ≥98.0% (by HPLC, total nitrogen)
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
Formula: C₁₂H₁₄N₂O₂·HCl
MW: 254,7 g/mol
Pto. de fusión: 205°C
Número MDL: MFCD00038992
Núm. CAS: 14907-27-8

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -17.0 to -22.0 deg (C=5, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR