L-Tyrosine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

T2443-25G T2443-5G
TCIAT2443-25GEA 39 EUR
TCIAT2443-25G TCIAT2443-5G
L-Tyrosine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)
L-Tyrosine benzyl ester-p-toluenesulphonate
Formula: C₁₆H₁₇NO₃
Número MDL: MFCD00038959
Núm. CAS: 53587-11-4
EINECS: 258-650-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C
Specific rotation [a]20/D -11.0 to -15.0 deg (C=3, MeOH)

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR