Tetraethyl-4-tert-butylcalix[4]arene-O,O',O'',O'''-tetraacetate ≥96.0% (by HPLC)
Proveedor: TCI
Sinónimos:
4-tert-Butylcalix[4]arene-O,O',O'',O'''-tetraacetic Acid Tetraethyl Ester, Tetraethyl 2,2',2'',2'''-((15,35,55,75-tetra-tert-butyl-1,3,5,7(1,3)-tetrabenzenacyclooctaphane-12,32,52,72-tetrayl)tetrakis(oxy))tetraacetate
T2563-1G
T2563-200MG
TCIAT2563-1GEA
397
EUR
TCIAT2563-1G
TCIAT2563-200MG
Tetraethyl-4-tert-butylcalix[4]arene-O,O',O'',O'''-tetraacetate ≥96.0% (by HPLC)
Tetraethyl-4-tert-butylcalix[4]arene-O,O',O'',O'''-tetraacetate
Formula:
C₆₀H₈₀O₁₂ MW: 993,29 g/mol |
Número MDL:
MFCD00145373 Núm. CAS: 97600-39-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Melting point | 152.0 to 157.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...