1,3,5-Tri-O-benzoyl-ɑ-D-ribofuranose ≥97.0% (by HPLC)

Proveedor: TCI

Sinónimos: 1,3,5-Tri-O-benzoyl-α-D-ribofuranose, 1,3,5-Tri-O-benzoyl-a-D-ribofuranose

T2641-25G T2641-5G
TCIAT2641-25GEA 208 EUR
TCIAT2641-25G TCIAT2641-5G
1,3,5-Tri-O-benzoyl-ɑ-D-ribofuranose ≥97.0% (by HPLC)
1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose
Formula: C₂₆H₂₂O₈
MW: 462,45 g/mol
Número MDL: MFCD00080818
Núm. CAS: 22224-41-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 138.0 to 142.0 deg-C
Specific rotation [a]20/D +82.0 to +88.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR