Tris(2-pyridylmethyl)amine ≥98.0% (by HPLC)

Proveedor: TCI
Atención

T2671-1G T2671-5G
TCIAT2671-1GEA 104 EUR
TCIAT2671-1G TCIAT2671-5G
Tris(2-pyridylmethyl)amine ≥98.0% (by HPLC)
Tris(2-pyridylmethyl)amine
Formula: C₁₈H₁₈N₄
MW: 290,37 g/mol
Número MDL: MFCD14708176
Núm. CAS: 16858-01-8

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR