Trimorpholinophosphine oxide ≥98.0% (por GC)

Proveedor: TCI

T2748-25G T2748-5G
TCIAT2748-25GEA 79 EUR
TCIAT2748-25G TCIAT2748-5G
Trimorpholinophosphine oxide ≥98.0% (por GC)
Trimorpholinophosphine oxide
Formula: C₁₂H₂₄N₃O₄P
Número MDL: MFCD00047406
Núm. CAS: 4441-12-7
EINECS: 224-662-7

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 189.0 to 193.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR