cis-1,2,3,6-Tetrahydrophthalimide ≥98.0% (by GC, titration analysis)

Proveedor: TCI

Sinónimos: cis-4-Cyclohexene-1,2-dicarboximide, cis-3a,4,7,7a-Tetrahydro-1H-isoindole-1,3(2H)-dione

T2801-250G T2801-25G
TCIAT2801-250GEA 83 EUR
TCIAT2801-250G TCIAT2801-25G
cis-1,2,3,6-Tetrahydrophthalimide ≥98.0% (by GC, titration analysis)
cis-1,2,3,6-Tetrahydrophthalimide
Formula: C₈H₉NO₂
MW: 151,17 g/mol
Número MDL: MFCD00005880
Núm. CAS: 1469-48-3
EINECS: 215-999-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=968aad4b59eec3df05633a743bb7c879177f5894&vid=968aad4b59eec3df05633a743bb7c879177f5894&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR