Thiamphenicol ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: 2,2-Dichloro-N-[(1R,2R)-1,3-dihydroxy-1-[4-(methylsulphonyl)phenyl]-2-propyl]acetamide, (Methylsulphonyl)chloramphenicol

T2802-5G T2802-25G
TCIAT2802-5GEA 52 EUR
TCIAT2802-5G TCIAT2802-25G
Thiamphenicol ≥98.0% (by HPLC, total nitrogen)
Thiamphenicol
Formula: C₁₂H₁₅Cl₂NO₅S
MW: 356,21 g/mol
Número MDL: MFCD00467983
Núm. CAS: 15318-45-3
EINECS: 239-355-3
Índice Merck: 14,09301

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 164.0 to 168.0 deg-C
Specific rotation [a]20/D +10.0 to +13.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR