N-(p-Toluenesulphonyl)glycine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
N-Tosylglycine, N-p-Tosylglycine, Tos-Gly-OH
T2803-25G
T2803-5G
TCIAT2803-25GEA
109
EUR
TCIAT2803-25G
TCIAT2803-5G
N-(p-Toluenesulphonyl)glycine ≥98.0% (by HPLC, titration analysis)
N-(p-Toluenesulphonyl)glycine
Formula:
C₉H₁₁NO₄S MW: 229,25 g/mol |
Número MDL:
MFCD00045898 Núm. CAS: 1080-44-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...