Tilorone dihydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2,7-Bis[2-(diethylamino)ethoxy]-9-fluorenone Dihydrochloride, 2,7-Bis(2-(Diethylamino)ethoxy)-9H-fluoren-9-one dihydrochloride

T2836-1G T2836-100MG
TCIAT2836-1GEA 201 EUR
TCIAT2836-1G TCIAT2836-100MG
Tilorone dihydrochloride ≥98.0% (by HPLC, titration analysis)
Tilorone dihydrochloride
Formula: C₂₅H₃₄N₂O₃
MW: 483,47 g/mol
Pto. de fusión: 234°C
Número MDL: MFCD00134071
Núm. CAS: 27591-69-1
Índice Merck: 14,09440

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR