N-(Triphenylmethyl)-L-serine methyl ester ≥98.0% (by GC, titration analysis)

Proveedor: TCI

Sinónimos: (S)-Methyl 3-hydroxy-2-(tritylamino)propanoate, Trt-Ser-OMe, N-Trityl-L-serine Methyl Ester

T2870-5G T2870-25G
TCIAT2870-5GEA 38 EUR
TCIAT2870-5G TCIAT2870-25G
N-(Triphenylmethyl)-L-serine methyl ester ≥98.0% (by GC, titration analysis)
N-(Triphenylmethyl)-L-serine methyl ester
Formula: C₂₃H₂₃NO₃
MW: 361,44 g/mol
Número MDL: MFCD00192570
Núm. CAS: 4465-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e38da92c3c2ced5514fa459d0a73b6cba95a9cc0&vid=e38da92c3c2ced5514fa459d0a73b6cba95a9cc0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C
Specific rotation [a]20/D +31.0 to +35.0 deg (C=1、MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR