Tris(3-metoxifenil)fosfina ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
T2946-25G
T2946-5G
TCIAT2946-25GEA
158
EUR
TCIAT2946-25G
TCIAT2946-5G
Tris(3-metoxifenil)fosfina ≥98.0% (by HPLC)
Tris(3-metoxifenil)fosfina
Formula:
C₂₁H₂₁O₃P MW: 352,37 g/mol |
Número MDL:
MFCD00008386 Núm. CAS: 29949-84-6 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Iodometric Titration) | min. 95.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...