6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
T2955-1G
T2955-5G
TCIAT2955-1GEA
26
EUR
TCIAT2955-1G
TCIAT2955-5G
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
Formula:
C₅H₆N₂O₃ |
Número MDL:
MFCD00052030 Núm. CAS: 27372-38-9 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...