5-(Trifluoromethyl)picolinic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 5-(Trifluoromethyl)pyridine-2-carboxylic acid

T2975-1G T2975-5G
TCIAT2975-1GEA 35 EUR
TCIAT2975-1G TCIAT2975-5G
5-(Trifluoromethyl)picolinic acid ≥98.0% (by GC, titration analysis)
5-(Trifluoromethyl)picolinic acid
Formula: C₇H₄F₃NO₂
MW: 191,11 g/mol
Número MDL: MFCD04113632
Núm. CAS: 80194-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb05de297f5a2499e809dfc87c063166571600e8&vid=fb05de297f5a2499e809dfc87c063166571600e8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 135.0 to 139.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR