Tiropramide hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

T2978-1G T2978-5G
TCIAT2978-1GEA 65 EUR
TCIAT2978-1G TCIAT2978-5G
Tiropramide hydrochloride ≥98.0% (by HPLC, titration analysis)
Tiropramide hydrochloride
Formula: C₂₈H₄₁N₃O₃
Número MDL: MFCD01741847
Núm. CAS: 53567-47-8
Índice Merck: 14,09466
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=018796ae01fa41213531e9f5fd4221926ebed0c1&vid=018796ae01fa41213531e9f5fd4221926ebed0c1&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 180.0 to 184.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR