Tanshinone IIA ≥97.0% (by HPLC)

Proveedor: TCI

T2987-10MG T2987-50MG
TCIAT2987-10MGEA 49 EUR
TCIAT2987-10MG TCIAT2987-50MG
Tanshinone IIA ≥97.0% (by HPLC)
Tanshinone IIA
Formula: C₁₉H₁₈O₃
Temperatura de almacenaje: Nevera
Número MDL: MFCD00238692
Núm. CAS: 568-72-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3926a6d2b29bd82b0984e3d19c5fae695d9f809&vid=c3926a6d2b29bd82b0984e3d19c5fae695d9f809&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 197.0 to 202.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR