1-Trifluoromethyl-1,2-benziodoxol-3(1H)-one (contains 60% Diatomaceous earth)

Proveedor: TCI
Peligro

Sinónimos: Togni Reagent II

T3014-1G T3014-5G
TCIAT3014-1GEA 44 EUR
TCIAT3014-1G TCIAT3014-5G
1-Trifluoromethyl-1,2-benziodoxol-3(1H)-one (contains 60% Diatomaceous earth)
1-Trifluoromethyl-1,2-benziodoxol-3(1H)-one
Formula: C₈H₄F₃IO₂
MW: 316,02 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD18800706
Núm. CAS: 887144-94-7
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=259e07d75b33e7803f34792229b51e64caad2ebf&vid=259e07d75b33e7803f34792229b51e64caad2ebf&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Assay 38 to 44 % (with CH2Cl2 insoluble matter)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR