Trimethyl-1,2,4-cyclohexanetricarboxylate (cis and trans mixture) cis- and trans-mixture ≥97.0% (por GC)
Proveedor: TCI
Sinónimos:
1,2,4-Cyclohexanetricarboxylic Acid Trimethyl Ester
T3029-1G
T3029-5G
TCIAT3029-1GEA
115
EUR
TCIAT3029-1G
TCIAT3029-5G
Trimethyl-1,2,4-cyclohexanetricarboxylate (cis and trans mixture) cis- and trans-mixture ≥97.0% (por GC)
Trimethyl-1,2,4-cyclohexanetricarboxylate (cis and trans mixture)
Formula:
C₁₂H₁₈O₆ MW: 258,27 g/mol Temperatura de almacenaje: Nevera |
Núm. CAS:
185855-30-5 |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % (total of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...