Trimebutine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 3,4,5-Trimethoxybenzoic Acid 2-(Dimethylamino)-2-phenylbutyl Ester, 2-(Dimethylamino)-2-phenylbutyl 3,4,5-Trimethoxybenzoate

T3049-5G T3049-25G
TCIAT3049-5GEA 44 EUR
TCIAT3049-5G TCIAT3049-25G
Trimebutine ≥98.0% (by HPLC, titration analysis)
Trimebutine
Formula: C₂₂H₂₉NO₅
MW: 387,48 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00133873
Núm. CAS: 39133-31-8
EINECS: 254-309-2
Índice Merck: 14,09699
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=442e4f470e1a1db2412ae71fa8c5d361af2bff51&vid=442e4f470e1a1db2412ae71fa8c5d361af2bff51&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 80.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR