2-(Trifluorometoxi)bencilamina ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: (2-(Trifluoromethoxy)phenyl)methanamine

T3092-1G T3092-5G
TCIAT3092-1GEA 41 EUR
TCIAT3092-1G TCIAT3092-5G
2-(Trifluorometoxi)bencilamina ≥98.0% (by GC, titration analysis)
2-(Trifluorometoxi)bencilamina
Formula: C₈H₈F₃NO
MW: 191,15 g/mol
Número MDL: MFCD00085176
Núm. CAS: 175205-64-8
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7a84ca5b5fa715760cc0fdaaa33e5212c4eed43&vid=b7a84ca5b5fa715760cc0fdaaa33e5212c4eed43&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow clear liquid to cloudy liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR