1H-1,2,4-Triazole-1-carboxamidine hydrochloride ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
1H-1,2,4-Triazole-1-carboximidamide hydrochloride
T3124-1G
T3124-5G
TCIAT3124-1GEA
16
EUR
TCIAT3124-1G
TCIAT3124-5G
1H-1,2,4-Triazole-1-carboxamidine hydrochloride ≥98.0% (by titrimetric analysis)
1H-1,2,4-Triazole-1-carboxamidine hydrochloride
Formula:
C₃H₆ClN₅ MW: 147,566 g/mol Pto. de fusión: 199°C |
Número MDL:
MFCD03095468 Núm. CAS: 19503-26-5 UN: 2811 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...