1-(Trifluoromethanesulphonyl)-1H-benzotriazole ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
T3167-1G
T3167-5G
TCIAT3167-1GEA
95
EUR
TCIAT3167-1G
TCIAT3167-5G
1-(Trifluoromethanesulphonyl)-1H-benzotriazole ≥98.0% (by HPLC)
1-(Trifluoromethanesulphonyl)-1H-benzotriazole
Formula:
C₇H₄F₃N₃O₂S MW: 251,18 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00963686 Núm. CAS: 117632-84-5 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 36.0 to 40.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...