S-p-Tolyl p-Toluenesulphonothioate ≥98.0% (by HPLC)

Proveedor: TCI

T3173-1G T3173-5G
TCIAT3173-1GEA 83 EUR
TCIAT3173-1G TCIAT3173-5G
S-p-Tolyl p-Toluenesulphonothioate ≥98.0% (by HPLC)
S-p-Tolyl p-Toluenesulphonothioate
Formula: C₁₄H₁₄O₂S₂
Número MDL: MFCD00092126
Núm. CAS: 2943-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fff144a1f14d8e6bf05880397356ede50bca5959&vid=fff144a1f14d8e6bf05880397356ede50bca5959&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 72.0 to 76.0 deg-C

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR