Tiaprofenic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

T3175-25MG T3175-100MG
TCIAT3175-25MGEA 67 EUR
TCIAT3175-25MG TCIAT3175-100MG
Tiaprofenic acid ≥98.0% (by GC, titration analysis)
Tiaprofenic acid
Formula: C₁₄H₁₂O₃S
Número MDL: MFCD00866089
Núm. CAS: 33005-95-7
EINECS: 251-329-3
UN: 2811
ADR: 6.1,III
Índice Merck: 14,09422
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30e2e9a3555639500570aa7da278bf498ad9b285&vid=30e2e9a3555639500570aa7da278bf498ad9b285&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR