2,4,6-Tris(4-bromophenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

T3178-200MG T3178-1G
TCIAT3178-200MGEA 57 EUR
TCIAT3178-200MG TCIAT3178-1G
2,4,6-Tris(4-bromophenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2,4,6-Tris(4-bromophenyl)-1,3,5-triazine
Formula: C₂₁H₁₂Br₃N₃
MW: 546,06 g/mol
Número MDL: MFCD04116312
Núm. CAS: 30363-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1b10d1c8ef0d3bf157a709ad88139cb30b7fcfb&vid=af0ea0e217d59d507cd85c3f4ce04babd0ed43ee&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR