1,3,5-Tri(1-naphthyl)benzene ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 1,3,5-Tri(naphthalen-1-yl)benzene

T3193-200MG T3193-1G
TCIAT3193-200MGEA 71 EUR
TCIAT3193-200MG TCIAT3193-1G
1,3,5-Tri(1-naphthyl)benzene ≥98.0% (by HPLC)
1,3,5-Tri(1-naphthyl)benzene
Formula: C₃₆H₂₄
Número MDL: MFCD28975119
Núm. CAS: 7059-70-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c907ce66e8698ec7f84c24a1ee6f34ffab7203&vid=b0c907ce66e8698ec7f84c24a1ee6f34ffab7203&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 184.0 to 188.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR