1-Naphthyl boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Naphthalene-1-boronic acid Pinacol ester, 1-Naphthalenboronic acid, pinacol ester, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene, 4,4,5,5-Tetramethyl-2-(1-naphthyl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborolane

T3218-1G T3218-5G
TCIAT3218-1GEA 29 EUR
TCIAT3218-1G TCIAT3218-5G
1-Naphthyl boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
1-Naphthyl boronic acid pinacol ester
Formula: C₁₆H₁₉BO₂
Número MDL: MFCD05663869
Núm. CAS: 68716-52-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50ae9005379f3db3676d6355fafcb8caa62765c1&vid=50ae9005379f3db3676d6355fafcb8caa62765c1&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR