4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate ≥96.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
T3224-1G
T3224-5G
TCIAT3224-1GEA
61
EUR
TCIAT3224-1G
TCIAT3224-5G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate ≥96.0% (by GC, titration analysis)
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate
Formula:
C₁₄H₁₉BO₄ |
Número MDL:
MFCD03453060 Núm. CAS: 480424-70-2 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 64.0 to 68.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...