Ácido 4'-(trifluorometil)bifenil-2-carboxílico ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-(Trifluoromethyl)-2'-biphenylcarboxylic acid, 4'-(Trifluoromethyl)-2-biphenylcarboxylic acid, 4'-Trifluoromethyl-2-biphenyl carboxylic acid, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-2-carboxylic acid
T3231-1G
T3231-5G
TCIAT3231-1GEA
24
EUR
TCIAT3231-1G
TCIAT3231-5G
Ácido 4'-(trifluorometil)bifenil-2-carboxílico ≥98.0% (by GC, titration analysis)
Ácido 4'-(trifluorometil)bifenil-2-carboxílico
Formula:
C₁₄H₉F₃O₂ MW: 266,22 g/mol |
Número MDL:
MFCD00075353 Núm. CAS: 84392-17-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 169.0 to 173.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...