4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzothiophene ≥98.0% (by GC, titration analysis)

Proveedor: TCI

Sinónimos: 2-(Dibenzo[b,d]thiophen-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(Dibenzothiophene-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Dibenzothiopheneboronic acid Pinacol ester

T3236-1G T3236-5G
TCIAT3236-1GEA 54 EUR
TCIAT3236-1G TCIAT3236-5G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzothiophene ≥98.0% (by GC, titration analysis)
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzothiophene
Formula: C₁₈H₁₉BO₂S
MW: 310,22 g/mol
Número MDL: MFCD18452170
Núm. CAS: 912824-84-1

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 68.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR