2-Bromo-6-nitrofenol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B2203-25G
B2203-5G
TCIAB2203-25GEA
120
EUR
TCIAB2203-25G
TCIAB2203-5G
2-Bromo-6-nitrofenol ≥98.0% (by GC, titration analysis)
2-Bromo-6-nitrofenol
Formula:
C₆H₄BrNO₃ MW: 218,01 g/mol |
Número MDL:
MFCD02683216 Núm. CAS: 13073-25-1 |
Resultados test especificaciones
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...