N,N,N',N'-Tetraphenylbenzidine ≥99.0% (por GC), purified by sublimation

Proveedor: TCI

Sinónimos: TPB, N,N,N,N-Tetraphenylbenzidine, 4,4'-Bis(diphenylamino)biphenyl

T3266-1G T3266-5G
TCIAT3266-1GEA 55 EUR
TCIAT3266-1G TCIAT3266-5G
N,N,N',N'-Tetraphenylbenzidine ≥99.0% (por GC), purified by sublimation
N,N,N',N'-Tetraphenylbenzidine
Formula: C₃₆H₂₈N₂
MW: 488,63 g/mol
Número MDL: MFCD00228123
Núm. CAS: 15546-43-7
EINECS: 239-599-0

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 231.0 to 236.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR