Trospium chloride ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
rel-(1R,3r,5S)-3-(2-Hydroxy-2,2-diphenylacetoxy)spiro[bicyclo[3.2.1]octane-8,1'-pyrrolidin]-1'-ium chloride
T3305-200MG
T3305-1G
TCIAT3305-200MGEA
65
EUR
TCIAT3305-200MG
TCIAT3305-1G
Trospium chloride ≥98.0% (by HPLC, total nitrogen)
Trospium chloride
Formula:
C₂₅H₃₀ClNO₃ MW: 427,97 g/mol |
Número MDL:
MFCD00865254 Núm. CAS: 10405-02-4 EINECS: 233-875-4 Índice Merck: 14,09786 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...