2,4,6-Tris(3-bromophenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

T3308-1G T3308-5G
TCIAT3308-1GEA 55 EUR
TCIAT3308-1G TCIAT3308-5G
2,4,6-Tris(3-bromophenyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2,4,6-Tris(3-bromophenyl)-1,3,5-triazine
Formula: C₂₁H₁₂Br₃N₃
MW: 546,06 g/mol
Número MDL: MFCD28964709
Núm. CAS: 890148-78-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 207.0 to 211.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR