2-(Trifluorometil)bencenosulfonamida ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
α,α,α-Trifluoro-o-toluenosulfonamida
T3368-5G
T3368-25G
TCIAT3368-5GEA
61
EUR
TCIAT3368-5G
TCIAT3368-25G
2-(Trifluorometil)bencenosulfonamida ≥98.0% (by HPLC, total nitrogen)
2-(Trifluorometil)bencenosulfonamida
Formula:
C₇H₆F₃NO₂S MW: 225,19 g/mol |
Número MDL:
MFCD00042420 Núm. CAS: 1869-24-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 189.0 to 193.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...