2,4,5-Triiodoimidazol ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: 2,4,5-Triiodo-1H-imidazol

T3369-1G T3369-5G
TCIAT3369-1GEA 32 EUR
TCIAT3369-1G TCIAT3369-5G
2,4,5-Triiodoimidazol ≥98.0% (by titrimetric analysis)
2,4,5-Triiodoimidazol
Formula: C₃HI₃N₂
MW: 445,77 g/mol
Número MDL: MFCD00068812
Núm. CAS: 1746-25-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a34e9b48b2ffa959144de32ada2b5e383589ad49&vid=a34e9b48b2ffa959144de32ada2b5e383589ad49&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 184.0 to 190.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR