1-(2-Metilfenil)piperazina ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
T3404-5G
T3404-25G
TCIAT3404-5GEA
38
EUR
TCIAT3404-5G
TCIAT3404-25G
1-(2-Metilfenil)piperazina ≥98.0% (by GC, titration analysis)
1-(2-Metilfenil)piperazina
Formula:
C₁₁H₁₆N₂ MW: 176.26 g/mol Pto. Ebullición: 161°C |
Número MDL:
MFCD00040729 Núm. CAS: 39512-51-1 EINECS: 254-481-9 UN: 3263 ADR: 8 III |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...