5-(2-Pyridyl)-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
2-(1H-Tetrazol-5-yl)pyridine, 5-(2-Pyridyl)-1H-tetrazole
T3431-1G
T3431-5G
TCIAT3431-1GEA
68
EUR
TCIAT3431-1G
TCIAT3431-5G
5-(2-Pyridyl)-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
5-(2-Pyridyl)-1H-tetrazole
Formula:
C₆H₅N₅ MW: 147,14 g/mol |
Número MDL:
MFCD00068114 Núm. CAS: 33893-89-9 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...