4-(9-Carbazolyl)benzeneboronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
9-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole, 9-Carbazolylbenzeneboronic acid pinacol ester
T3452-1G
T3452-5G
TCIAT3452-1GEA
39
EUR
TCIAT3452-1G
TCIAT3452-5G
4-(9-Carbazolyl)benzeneboronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
4-(9-Carbazolyl)benzeneboronic acid pinacol ester
Formula:
C₂₄H₂₄BNO₂ MW: 369,27 g/mol |
Número MDL:
MFCD16294549 Núm. CAS: 785051-54-9 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 176.0 to 180.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...