1,2,3-Trifluoro-5-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene ≥98.0% (por GC)
Proveedor: TCI
T3466-1G
T3466-5G
TCIAT3466-1GEA
92
EUR
TCIAT3466-1G
TCIAT3466-5G
1,2,3-Trifluoro-5-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene ≥98.0% (por GC)
1,2,3-Trifluoro-5-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene
Formula:
C₂₃H₃₃F₃ MW: 366,51 g/mol |
Número MDL:
MFCD12828865 Núm. CAS: 131819-24-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...