4,4,5,5-Tetramethyl-2-[4-[(tetrahydro-2H-pyran-2-yl)oxy]phenyl]-1,3,2-dioxaborolane ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolane-2-yl)phenoxy]tetrahydro-2H-pyran, 4,4,5,5-Tetramethyl-2-(4-((tetrahydro-2H-pyran-2-yl)oxy)phenyl)-1,3,2-dioxaborolane, 4-[(Tetrahydro-2H-pyran-2-yl)oxy]phenylboronic acid Pinacol ester
T3468-1G
T3468-5G
TCIAT3468-1GEA
99
EUR
TCIAT3468-1G
TCIAT3468-5G
4,4,5,5-Tetramethyl-2-[4-[(tetrahydro-2H-pyran-2-yl)oxy]phenyl]-1,3,2-dioxaborolane ≥98.0% (by titrimetric analysis)
4,4,5,5-Tetramethyl-2-[4-[(tetrahydro-2H-pyran-2-yl)oxy]phenyl]-1,3,2-dioxaborolane
Formula:
C₁₇H₂₅BO₄ MW: 304,19 g/mol |
Número MDL:
MFCD06659834 Núm. CAS: 889865-38-7 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 57.0 to 61.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...