1,3,5-Tris[(1H-imidazol-1-yl)methyl]benzene ≥95.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
1,3,5-Tris((1H-imidazol-1-yl)methyl)benzene
T3479-1G
T3479-5G
TCIAT3479-1GEA
73
EUR
TCIAT3479-1G
TCIAT3479-5G
1,3,5-Tris[(1H-imidazol-1-yl)methyl]benzene ≥95.0% (by HPLC, titration analysis)
1,3,5-Tris[(1H-imidazol-1-yl)methyl]benzene
Formula:
C₁₈H₁₈N₆ MW: 318,38 g/mol |
Número MDL:
MFCD32062936 Núm. CAS: 147951-02-8 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 183.0 to 187.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...