5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
5-Formyl-2-thiopheneboronic acid Pinacol ester, 2-(5-Formyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde
T3485-1G
T3485-5G
TCIAT3485-1GEA
32
EUR
TCIAT3485-1G
TCIAT3485-5G
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde ≥98.0% (by GC, titration analysis)
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde
Formula:
C₁₁H₁₅BO₃S MW: 238,11 g/mol |
Número MDL:
MFCD09266186 Núm. CAS: 1040281-83-1 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...